Learn More
Medchemexpress LLC FGF-21 Human CHO 100ug
SDP
Catalog No. 5000180344
Encompass_Preferred
FGF-21 Protein Human (CHO His) is a polypeptide chain containing the C-termimal His tag produced in CHO cells FGF-21 Protein Human (CHO His) is a member of the broblast growth factor (FGF) subfamily acts as a metabolic regulator with pleiotropic effects
Catalog No. 50-001-80344 Supplier Medchemexpress LLC Supplier No. HYP7345100UG
FGF-21 Protein Human (CHO His) is a polypeptide chain containing the C-termimal His tag produced in CHO cells FGF-21 Protein Human (CHO His) is a member of the broblast growth factor (FGF) subfamily acts as a metabolic regulator with pleiotropic effects