Learn More
Medchemexpress LLC FGF-21 Mouse HEK29 500ug
SDP
Catalog No. 5000182850
Encompass_Preferred
FGF-21 Proteinas influences glucose uptake in adipocytes by inducing SLC2A1/GLUT1 expression particularly with KLB s presence It plays a crucial role in systemic glucose homeostasis and insulin sensitivity interacting directly with KLB through its C-terminus and engaging with FGFR4 The protein s molecular mechanisms involve a complex interplay emphasizing its broad impact beyond localized effects FGF-21 Protein Mouse (HEK293 His) is the recombinant mouse-derived FGF-21 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82850 Supplier Medchemexpress LLC Supplier No. HYP72651500UG
FGF-21 Proteinas influences glucose uptake in adipocytes by inducing SLC2A1/GLUT1 expression particularly with KLB s presence It plays a crucial role in systemic glucose homeostasis and insulin sensitivity interacting directly with KLB through its C-terminus and engaging with FGFR4 The protein s molecular mechanisms involve a complex interplay emphasizing its broad impact beyond localized effects FGF-21 Protein Mouse (HEK293 His) is the recombinant mouse-derived FGF-21 protein expressed by HEK293 with C-6 His labeled tag