Learn More
AdipoGen FGF-21 mouse pAb
SDP
Supplier:  AdipoGen AG25A0076C100
Encompass_Preferred
Polyclonal Antibody. Recognizes mouse FGF-21. Detects a band of ~25kDa by Western blot. Weakly cross-reacts with human FGF-21. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. FGF-21 stimulates glucose uptake in differentiated adipocytes via the induction of glucose transporter SLC2A1/GLUT1 expression but not SLC2A4/GLUT4 expression. Activity requires the presence of bKlotho.
Catalog No. 50-256-3239