Learn More
AdipoGen FGF-21 (mouse) (rec.) (10 µg)
SDP
Supplier:  AdipoGen AG40B0143C010
Encompass_Preferred
Protein. Mouse FGF-21 (aa 29-210) is fused at the N-terminus to a FLAG-tag. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). FGF-21 is a member of the fibroblast growth factor family and has been identified as an important regulator of energy metabolism connecting nutrition, growth, reproduction and longevity. In adipose tissue, FGF-21 promotes glucose uptake and oxidation and in liver it replenishes on fasting the tissues with fuel on low nutritional supply. FGF-21 acts through FGF receptors (FGFR) associated with the auxiliary protein beta-Klotho. Recently, FGF-21 has been reported to interact directly with the brain circadian clock to coordinate activity and reproduction as part of the adaptation to fasting. Due to its multiple functions of normalizing glucose, lipid and energy homeostasis, FGF-21 represents an attractive novel therapy for type 2 diabetes mellitus and obesity.
Catalog No. 50-205-3228