Learn More
Medchemexpress LLC FGF-21 Protein, Mouse (HEK293, His) | ≥ 95% | 20.8 kDa (predicted); 20-25 kDa (SDS-PAGE) | 1 MG
SDP
Catalog No. 5000399195
Encompass_Preferred
FGF-21 Protein, Mouse (HEK293, His) influences glucose uptake in adipocytes by inducing SLC2A1/GLUT1 expression, particularly with KLB's presence. It plays a crucial role in systemic glucose homeostasis and insulin sensitivity, interacting directly with KLB through its C-terminus and engaging with FGFR4. The protein's molecular mechanisms involve a complex interplay, emphasizing its broad impact beyond localized effects. It is the recombinant mouse-derived FGF-21 protein, expressed by HEK293, with a C-6*His labeled tag.
  • Influences glucose uptake in adipocytes by inducing SLC2A1/GLUT1 expression.
  • Plays a crucial role in systemic glucose homeostasis and insulin sensitivity.
  • Interacts directly with KLB through its C-terminus and engages with FGFR4.
  • Recombinant mouse-derived protein.
  • Expressed by HEK293.
  • Features a C-6*His labeled tag.
Catalog No. 50-003-99195 Supplier Medchemexpress LLC Supplier No. HYP726511MG
FGF-21 Protein, Mouse (HEK293, His) influences glucose uptake in adipocytes by inducing SLC2A1/GLUT1 expression, particularly with KLB's presence. It plays a crucial role in systemic glucose homeostasis and insulin sensitivity, interacting directly with KLB through its C-terminus and engaging with FGFR4. The protein's molecular mechanisms involve a complex interplay, emphasizing its broad impact beyond localized effects. It is the recombinant mouse-derived FGF-21 protein, expressed by HEK293, with a C-6*His labeled tag.
  • Influences glucose uptake in adipocytes by inducing SLC2A1/GLUT1 expression.
  • Plays a crucial role in systemic glucose homeostasis and insulin sensitivity.
  • Interacts directly with KLB through its C-terminus and engages with FGFR4.
  • Recombinant mouse-derived protein.
  • Expressed by HEK293.
  • Features a C-6*His labeled tag.