Learn More
Medchemexpress LLC Fibroblast growth factor 22 | 93.0% | 50UG
SDP
Catalog No. 5000176551
Encompass_Preferred
FGF-22 Protein, Human (His-SUMO) is a recombinant human fibroblast growth factor produced for in vitro research and cell-culture applications. The protein is expressed in Escherichia coli and carries an N-terminal SUMO fusion and an N-terminal His tag to facilitate purification and handling. FGF-22 is involved in FGF signaling and has roles in metabolic regulation, cell proliferation, and tissue development.
  • Recombinant human FGF-22 sequence suitable for in vitro studies.
  • N-terminal SUMO and His tags enable efficient purification.
  • Expressed in Escherichia coli for scalable recombinant production.
  • Suitable for cell culture and biochemical assays.
  • Relevant to studies of metabolic regulation and tissue development.
Catalog No. 50-001-76551 Supplier Medchemexpress LLC Supplier No. HYP70049450UG
FGF-22 Protein, Human (His-SUMO) is a recombinant human fibroblast growth factor produced for in vitro research and cell-culture applications. The protein is expressed in Escherichia coli and carries an N-terminal SUMO fusion and an N-terminal His tag to facilitate purification and handling. FGF-22 is involved in FGF signaling and has roles in metabolic regulation, cell proliferation, and tissue development.
  • Recombinant human FGF-22 sequence suitable for in vitro studies.
  • N-terminal SUMO and His tags enable efficient purification.
  • Expressed in Escherichia coli for scalable recombinant production.
  • Suitable for cell culture and biochemical assays.
  • Relevant to studies of metabolic regulation and tissue development.