Learn More
BioVendor, LLC Fgf-23 Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL007R
Encompass
Fibroblast growth factor-23 (FGF-23) is a phosphaturic hormone involved in mineral bone metabolism that helps control phosphate homeostasis and reduces 1,25- dihydroxyvitamin D synthesis. Recent data have highlighted the relevant direct FGF-23 effects on the myocardium, and high plasma levels of FGF-23 have been associated with adverse cardiovascular outcomes in humans, such as heart failure and arrhythmias. Therefore, FGF-23 has emerged as a novel biomarker of cardiovascular risk. experimental data suggest FGF-23 as a direct mediator of cardiac hypertrophy development, cardiac fibrosis and cardiac dysfunction via specific myocardial FGF receptor (FGFR) activation. One kit (96 wells). Type: sandwich ELISA. Sample type: Plasma (EDTA), urine. Sample requirement: 50 ¿l/well. Calibration range: 50-2000pg/ml. Limit of detection: 25pg/ml. Other names: Fibroblast Growth Factor 23, Phosphatonin, Tumor-derived hypophosphatemia-inducing factor, FGF23.
Catalog No. 50-240-6358