Learn More
AdipoGen FGF-23 human rec His
SDP
Catalog No. 502562991
Encompass_Preferred
Protein. Signal peptide and human FGF-23 aa 1-251 are fused at the C-terminus to a His-tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >90% SDS-PAGE. FGF-23 Fibroblast growth factor 23 is a regulator of phosphate homeostasis. It upregulates EGR1 expression in the presence of KLBy. Acts directly on the parathyroid to decrease PTH secretion. Regulates the vitamin-D metabolism. Negatively regulates osteoblast differentiation and matrix mineralization. Defects in FGF-23 are the cause of autosomal dominant hypophosphataemic rickets ADHR and of hyperphosphatemic familial tumoral calcinosis HFTC.
Catalog No. 50-256-2991 Supplier AdipoGen Supplier No. AG40A0114YC010
Protein. Signal peptide and human FGF-23 aa 1-251 are fused at the C-terminus to a His-tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >90% SDS-PAGE. FGF-23 Fibroblast growth factor 23 is a regulator of phosphate homeostasis. It upregulates EGR1 expression in the presence of KLBy. Acts directly on the parathyroid to decrease PTH secretion. Regulates the vitamin-D metabolism. Negatively regulates osteoblast differentiation and matrix mineralization. Defects in FGF-23 are the cause of autosomal dominant hypophosphataemic rickets ADHR and of hyperphosphatemic familial tumoral calcinosis HFTC.