Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGF-4 Human 153a.a 50ug
SDP

Catalog No. 5000184068
Encompass_Preferred

Recombinant human fibroblast growth factor 4 (FGF-4) is a 153-amino-acid, lyophilized protein used in research to study embryonic development, cell proliferation, and differentiation. The protein is provided in a stabilized formulation to preserve activity and is intended for in vitro signaling and developmental biology applications.

  • Human sequence of 153 amino acids.
  • Approximately 16.0 kDa molecular weight.
  • Lyophilized formulation for improved shelf life.
  • Stable storage at -20°C; aliquot and store at -80°C for extended preservation.
  • Contains formulation components such as buffer, sucrose, mannitol, and Tween-80.
  • UniProt accession P08620-1 corresponds to the provided sequence region.

Catalog No. 50-001-84068 Supplier Medchemexpress LLC Supplier No. HYP7069450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human fibroblast growth factor 4 (FGF-4) is a 153-amino-acid, lyophilized protein used in research to study embryonic development, cell proliferation, and differentiation. The protein is provided in a stabilized formulation to preserve activity and is intended for in vitro signaling and developmental biology applications.

  • Human sequence of 153 amino acids.
  • Approximately 16.0 kDa molecular weight.
  • Lyophilized formulation for improved shelf life.
  • Stable storage at -20°C; aliquot and store at -80°C for extended preservation.
  • Contains formulation components such as buffer, sucrose, mannitol, and Tween-80.
  • UniProt accession P08620-1 corresponds to the provided sequence region.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.