Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human fibroblast growth factor 4 | >95.0% | 50UG
SDP

Catalog No. 5000175243
Encompass_Preferred

Recombinant human fibroblast growth factor 4 (FGF-4) is a tag-free, E. coli-expressed protein used to stimulate cell proliferation and signaling in vitro. Supplied lyophilized and available in multiple pack sizes including a 50 μg option, it is intended for research use in stem cell maintenance, developmental biology, and related assays.

  • Expressed in E. coli, tag-free.
  • Purity >95.0% by reducing SDS-PAGE.
  • Predicted molecular mass 19.8 kDa; apparent 19-24 kDa by SDS-PAGE.
  • Lyophilized from HEPES/NaCl solution; trehalose option available.
  • High specific activity: ED50 ≤1.5 ng/mL in 3T3 assay.
  • Store lyophilized at -20°C; aliquot after reconstitution for long-term storage.

Catalog No. 50-001-75243 Supplier Medchemexpress LLC Supplier No. HYP7014A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human fibroblast growth factor 4 (FGF-4) is a tag-free, E. coli-expressed protein used to stimulate cell proliferation and signaling in vitro. Supplied lyophilized and available in multiple pack sizes including a 50 μg option, it is intended for research use in stem cell maintenance, developmental biology, and related assays.

  • Expressed in E. coli, tag-free.
  • Purity >95.0% by reducing SDS-PAGE.
  • Predicted molecular mass 19.8 kDa; apparent 19-24 kDa by SDS-PAGE.
  • Lyophilized from HEPES/NaCl solution; trehalose option available.
  • High specific activity: ED50 ≤1.5 ng/mL in 3T3 assay.
  • Store lyophilized at -20°C; aliquot after reconstitution for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.