Learn More
Genscript Corporation Antibody Protein Proteins Proteins 5 UG 1/EA
Supplier:  Genscript Corporation Z029845
Encompass
Fibroblast Growth Factor-4 (FGF-4) also known as K-FGF is a heparin-binding growth factor of the FGF family.It was identified by its oncogenic transforming activity. FGF-4 and FGF-3 are located closely on chromosome 11. FGF-4 and its receptors (FGF R1c 2c 3c and 4) play an important role in the regulation of embryonic development cell proliferation and cell differentiation. FGF-4 is required for normal limb and cardiac valve development during embryogenesis.
Catalog No. 50-252-9919