Learn More
Medchemexpress LLC FGF-4 Mouse 10ug
SDP
Catalog No. 5000182623
Encompass_Preferred
The FGF-4 protein plays a key role in embryonic development and is central to cell proliferation and differentiation It is essential for the survival of mouse embryos after implantation and is key to normal limb and heart valve development FGF-4 Protein Mouse is the recombinant mouse-derived FGF-4 protein expressed by E coli with tag free
Catalog No. 50-001-82623 Supplier Medchemexpress LLC Supplier No. HYP7264910UG
The FGF-4 protein plays a key role in embryonic development and is central to cell proliferation and differentiation It is essential for the survival of mouse embryos after implantation and is key to normal limb and heart valve development FGF-4 Protein Mouse is the recombinant mouse-derived FGF-4 protein expressed by E coli with tag free