Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGF-4 Protein, Human | ≥ 95% | 19-24 kDa | 250 UG
SDP

Catalog No. 5000396713
Encompass_Preferred

FGF-4 Protein is a recombinant protein expressed by *E. coli* without a tag. It is a member of the fibroblast growth factor family, activating RAS-MAPK and PI3K-AKT pathways by binding to FGFRs (specifically FGFR1 and FGFR2). This protein is crucial in physiological processes like cell growth, differentiation, migration, and angiogenesis, and is intended for research use only.

  • Activates RAS-MAPK and PI3K-AKT signaling pathways
  • Plays a crucial role in cell growth, differentiation, migration, and angiogenesis
  • Recombinant protein produced in *E. coli*
  • Purity of ≥ 95%

Catalog No. 50-003-96713 Supplier Medchemexpress LLC Supplier No. 250UGHYP7014
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FGF-4 Protein is a recombinant protein expressed by *E. coli* without a tag. It is a member of the fibroblast growth factor family, activating RAS-MAPK and PI3K-AKT pathways by binding to FGFRs (specifically FGFR1 and FGFR2). This protein is crucial in physiological processes like cell growth, differentiation, migration, and angiogenesis, and is intended for research use only.

  • Activates RAS-MAPK and PI3K-AKT signaling pathways
  • Plays a crucial role in cell growth, differentiation, migration, and angiogenesis
  • Recombinant protein produced in *E. coli*
  • Purity of ≥ 95%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.