Learn More
Enzo Life Sciences FGF-5 (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2310050
Encompass_Preferred
Alternative name: Fibroblast growth factor 5, Heparin-binding growth factor 5, HBGF-5, Smag-82. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 10 mM sodium chloride, pH 7.5. Source: Produced in E. coli. Contains 252 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4027