Learn More
Medchemexpress LLC HY-P7176 10ug Medchemexpress, FGF-7, Mouse CAS: Purity:>98%
SDP
Catalog No. 502083120
Encompass_Preferred
Medchemexpress, HY-P7176 10ug FGF-7, Mouse CAS: Recombinant Mouse Fibroblast Growth Factor-7 is a potent mitogen that enhances cell proliferation in various organs, including the skin, intestine, breast, liver, and lung. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3120 Supplier Medchemexpress LLC Supplier No. HYP717610UG
Medchemexpress, HY-P7176 10ug FGF-7, Mouse CAS: Recombinant Mouse Fibroblast Growth Factor-7 is a potent mitogen that enhances cell proliferation in various organs, including the skin, intestine, breast, liver, and lung. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.