Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGF-8b Human/Mouse 100ug
SDP

Catalog No. 5000185488
Encompass_Preferred

FGF-8b Protein, Human/Mouse is a recombinant, tag-free isoform of fibroblast growth factor 8 produced in Escherichia coli and supplied lyophilized for research applications. The product is characterized by provided amino acid sequence, purity data, and recommended formulation and storage instructions to support cell signaling and developmental biology studies.

  • Expressed in Escherichia coli for recombinant production.
  • Tag free to minimize interference in bioassays.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Provided lyophilized with recommended formulation options for stability.
  • Storage and reconstitution guidance included to preserve activity.

Catalog No. 50-001-85488 Supplier Medchemexpress LLC Supplier No. HYP70533100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FGF-8b Protein, Human/Mouse is a recombinant, tag-free isoform of fibroblast growth factor 8 produced in Escherichia coli and supplied lyophilized for research applications. The product is characterized by provided amino acid sequence, purity data, and recommended formulation and storage instructions to support cell signaling and developmental biology studies.

  • Expressed in Escherichia coli for recombinant production.
  • Tag free to minimize interference in bioassays.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Provided lyophilized with recommended formulation options for stability.
  • Storage and reconstitution guidance included to preserve activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.