Learn More
Medchemexpress LLC FGF-8e Human 193a. 100ug
SDP
Catalog No. 5000180666
Encompass_Preferred
FGF-8e Protein Human is an isoform of FGF-8 an essential factor during embryonic development involved in the progression of prostate cancer
Catalog No. 50-001-80666 Supplier Medchemexpress LLC Supplier No. HYP7346A100UG
FGF-8e Protein Human is an isoform of FGF-8 an essential factor during embryonic development involved in the progression of prostate cancer