Learn More
Medchemexpress LLC FGF-9 Human HEK293 2ug
SDP
Catalog No. 5000180500
Encompass_Preferred
Activin AB protein is a member of the transforming growth factor- (TGF- ) superfamily and plays a key role in various biological processes such as cell differentiation proliferation and apoptosis It is involved in embryonic development tissue repair and immune regulation FGF-9 Protein Human (HEK293 Fc solution) is the recombinant human-derived FGF-9 protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-80500 Supplier Medchemexpress LLC Supplier No. HYP730532UG
Activin AB protein is a member of the transforming growth factor- (TGF- ) superfamily and plays a key role in various biological processes such as cell differentiation proliferation and apoptosis It is involved in embryonic development tissue repair and immune regulation FGF-9 Protein Human (HEK293 Fc solution) is the recombinant human-derived FGF-9 protein expressed by HEK293 with N-hFc labeled tag