Learn More
Genscript Corporation Antibody Protein Proteins Proteins 100 UG 1/EA
Supplier:  Genscript Corporation Z03575100
Encompass
Fibroblast growth factor-9 (FGF-9) is an approximately 26 kDa secreted glycoprotein of the FGF family. Secreted mouse FGF-9 lacks the N-terminal 1-3 aa and shares greater than98% sequence identity with rat human equine porcine and bovine FGF-9.
Catalog No. 50-252-9615