Learn More
Medchemexpress LLC FGF-9 Mouse N-His 50ug
SDP
Catalog No. 5000180681
Encompass_Preferred
FGF-9 is a member of mouse heparin-binding fibroblast growth factors located on the outside of cell membranes FGF-9 is a regulator of chondrogenesis and vascularization during bone development FGF-9 binds to fibroblast growth factor receptors (FGFRs) to activate downstream signaling pathways such as PI3K-AKT RAS-MAPK and protein kinase C (PKC) pathways FGF-9 regulates embryonic development cell proliferation cell differentiation and cell migration FGF-9 plays an important role in differentiation survival of neuronal cells and growth stimulation of glial tumors FGF-9 Protein Mouse (N-His) is a recombinant protein with a N-His label that consisting of 208 amino acids and is produced in E coli[1][2][3][4][5]
Catalog No. 50-001-80681 Supplier Medchemexpress LLC Supplier No. HYP7352A50UG
FGF-9 is a member of mouse heparin-binding fibroblast growth factors located on the outside of cell membranes FGF-9 is a regulator of chondrogenesis and vascularization during bone development FGF-9 binds to fibroblast growth factor receptors (FGFRs) to activate downstream signaling pathways such as PI3K-AKT RAS-MAPK and protein kinase C (PKC) pathways FGF-9 regulates embryonic development cell proliferation cell differentiation and cell migration FGF-9 plays an important role in differentiation survival of neuronal cells and growth stimulation of glial tumors FGF-9 Protein Mouse (N-His) is a recombinant protein with a N-His label that consisting of 208 amino acids and is produced in E coli[1][2][3][4][5]