Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fgf basic/bfgf protein, human (P. pastoris, N-His) | >90.0% | 100UG
SDP

Supplier:  Medchemexpress LLC HYP700479100UG

Encompass_Preferred

Recombinant human basic fibroblast growth factor (bFGF/FGF-2) produced in Pichia pastoris and supplied as a purified, N-terminally His-tagged protein for research use. It is designed for cell biology, signaling assays, and biochemical characterization; manufacturer documentation provides purity, molecular weight, and storage guidance.

  • Recombinant human basic fibroblast growth factor for research use.
  • Expressed in Pichia pastoris for eukaryotic processing.
  • N-terminal 6xHis tag for purification and detection.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Approximate molecular weight ~20 kDa.
  • Available in small microgram formats suitable for lab assays.
  • Store at -20 °C; aliquot and freeze for long-term storage.

Catalog No. 50-001-76979


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.