Learn More
Medchemexpress LLC FGF basic/bFGF Huma 10ug
SDP
Catalog No. 5000179939
Encompass_Preferred
FGF-2 is a member of the fibroblast family involved in bone healing cartilage repair bone repair and nerve regeneration FGF-2 is also a mitotic promoter that accelerates cell proliferation FGF-2 regulates immune processes by specifically targeting tyrosine kinase receptors and activating the FGF/FGFR signaling pathway For example FGF-2 is involved in the JAK-STAT signaling pathway to regulate cartilage metabolism and also activates ERK signaling to promote cartilage regeneration FGF-2 combined with FGFR1/3 promoted degeneration and repair of articular cartilage respectively[1] FGF-2 is also a known carcinogen in GBM which contributes to glioma growth and vascularization[2] FGF basic/bFGF Protein Human (154a a His) consists of 154 amino acids produced by E coli with tag free
Catalog No. 50-001-79939 Supplier Medchemexpress LLC Supplier No. HYP7331A10UG
FGF-2 is a member of the fibroblast family involved in bone healing cartilage repair bone repair and nerve regeneration FGF-2 is also a mitotic promoter that accelerates cell proliferation FGF-2 regulates immune processes by specifically targeting tyrosine kinase receptors and activating the FGF/FGFR signaling pathway For example FGF-2 is involved in the JAK-STAT signaling pathway to regulate cartilage metabolism and also activates ERK signaling to promote cartilage regeneration FGF-2 combined with FGFR1/3 promoted degeneration and repair of articular cartilage respectively[1] FGF-2 is also a known carcinogen in GBM which contributes to glioma growth and vascularization[2] FGF basic/bFGF Protein Human (154a a His) consists of 154 amino acids produced by E coli with tag free