Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGF basic/bFGF Huma 1mg
SDP

Catalog No. 5000185541
Encompass_Preferred

Recombinant human basic fibroblast growth factor (bFGF/FGF-2) K128N, tag-free, supplied as a lyophilized powder for research use. The protein promotes cell proliferation and supports cartilage, bone, and nerve repair by activating FGF/FGFR signaling; bioactivity is validated in a cell proliferation assay.

  • Full-length human sequence (M1-S155) with K128N substitution.
  • E. coli-expressed, tag free recombinant protein.
  • Purity >95% by reducing SDS-PAGE.
  • Lyophilized formulation with stabilizers for storage.
  • Specific activity ≥1.89x10^6 units/mg (ED50 ≤0.53 ng/mL).
  • Stable at -20°C for long-term storage; aliquot to avoid freeze-thaw.

Catalog No. 50-001-85541 Supplier Medchemexpress LLC Supplier No. HYP70600A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human basic fibroblast growth factor (bFGF/FGF-2) K128N, tag-free, supplied as a lyophilized powder for research use. The protein promotes cell proliferation and supports cartilage, bone, and nerve repair by activating FGF/FGFR signaling; bioactivity is validated in a cell proliferation assay.

  • Full-length human sequence (M1-S155) with K128N substitution.
  • E. coli-expressed, tag free recombinant protein.
  • Purity >95% by reducing SDS-PAGE.
  • Lyophilized formulation with stabilizers for storage.
  • Specific activity ≥1.89x10^6 units/mg (ED50 ≤0.53 ng/mL).
  • Stable at -20°C for long-term storage; aliquot to avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.