Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fibroblast growth factor basic (bFGF), human | 106096-93-9 | MFCD00081382 | >95.0% | 500UG
SDP

Catalog No. 5000177695
Encompass_Preferred

Recombinant human basic fibroblast growth factor (bFGF/FGF-2), 146 amino acids, supplied as lyophilized recombinant protein for research applications. The product is tag-free, expressed in E. coli, and intended for cell biology and biochemical assays involving FGF signaling. Predicted mass is 16.4 kDa and purity is greater than 95% by reducing SDS-PAGE; multiple formulation options are available and long-term storage at -20°C or colder is recommended.

  • Recombinant human protein, 146 amino acids.
  • Tag free and expressed in E. coli.
  • Predicted molecular weight 16.4 kDa (SDS-PAGE ~14-20 kDa).
  • Purity >95% as determined by reducing SDS-PAGE.
  • Provided as lyophilized powder with formulation options.
  • Stable when stored at -20°C; freeze aliquots for long-term storage.
  • Suitable for cell culture and biochemical assays involving FGF signaling.

Catalog No. 50-001-77695 Supplier Medchemexpress LLC Supplier No. HYP7004500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human basic fibroblast growth factor (bFGF/FGF-2), 146 amino acids, supplied as lyophilized recombinant protein for research applications. The product is tag-free, expressed in E. coli, and intended for cell biology and biochemical assays involving FGF signaling. Predicted mass is 16.4 kDa and purity is greater than 95% by reducing SDS-PAGE; multiple formulation options are available and long-term storage at -20°C or colder is recommended.

  • Recombinant human protein, 146 amino acids.
  • Tag free and expressed in E. coli.
  • Predicted molecular weight 16.4 kDa (SDS-PAGE ~14-20 kDa).
  • Purity >95% as determined by reducing SDS-PAGE.
  • Provided as lyophilized powder with formulation options.
  • Stable when stored at -20°C; freeze aliquots for long-term storage.
  • Suitable for cell culture and biochemical assays involving FGF signaling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.