Learn More
Medchemexpress LLC Fibroblast growth factor 2 (bFGF) protein, human | >90.0% | 50UG
SDP
Catalog No. 5000175621
Encompass_Preferred
Recombinant human basic fibroblast growth factor (bFGF/FGF2) expressed in Pichia pastoris and supplied as an N-terminal 6xHis-tagged mature isoform. The product is provided lyophilized for research applications such as cell culture, proliferation assays, and signaling studies.
  • Expressed in Pichia pastoris with N-terminal 6xHis tag for detection and purification.
  • Mature protein corresponding to residues P143-S288; predicted mass 17.9 kDa.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Lyophilized formulation containing trehalose for enhanced stability.
  • Recommended reconstitution at ≥100 μg/mL; aliquot and store at -20°C or -80°C for long term.
  • Suitable for cell culture, biochemical assays, and growth factor signaling studies.
Catalog No. 50-001-75621 Supplier Medchemexpress LLC Supplier No. HYP70047950UG
Recombinant human basic fibroblast growth factor (bFGF/FGF2) expressed in Pichia pastoris and supplied as an N-terminal 6xHis-tagged mature isoform. The product is provided lyophilized for research applications such as cell culture, proliferation assays, and signaling studies.
  • Expressed in Pichia pastoris with N-terminal 6xHis tag for detection and purification.
  • Mature protein corresponding to residues P143-S288; predicted mass 17.9 kDa.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Lyophilized formulation containing trehalose for enhanced stability.
  • Recommended reconstitution at ≥100 μg/mL; aliquot and store at -20°C or -80°C for long term.
  • Suitable for cell culture, biochemical assays, and growth factor signaling studies.