Learn More
Medchemexpress LLC FGF basic/bFGF Protein, Human (146a.a) | >=98.0% | Approximately 14-20 kDa (SDS-PAGE), predicted 16.4 kDa | 250 UG
SDP
Catalog No. 5000396585
Encompass_Preferred
FGF basic/bFGF Protein, Human (146a.a) is a 146 amino acid protein produced by E.coli with tag-free purification. It plays a key role in bone healing, cartilage repair, bone repair, and nerve regeneration. This protein acts as a mitotic promoter, accelerating cell proliferation, and regulates immune processes by activating the FGF/FGFR signaling pathway, including the JAK-STAT and ERK pathways, to promote cartilage regeneration.
  • Supports tissue repair processes such as bone healing and nerve regeneration.
  • Promotes cell proliferation, differentiation, and apoptosis.
  • Activates FGF/FR signaling pathways to regulate cartilage metabolism.
  • Exhibits high biological activity with an ED50 of <2 ng/mL.
  • Implicated in cancer research, particularly in glioblastoma multiforme (GBM).
  • Produced in E. coli with tag-free purification.
Catalog No. 50-003-96585 Supplier Medchemexpress LLC Supplier No. 250UGHYP7004
FGF basic/bFGF Protein, Human (146a.a) is a 146 amino acid protein produced by E.coli with tag-free purification. It plays a key role in bone healing, cartilage repair, bone repair, and nerve regeneration. This protein acts as a mitotic promoter, accelerating cell proliferation, and regulates immune processes by activating the FGF/FGFR signaling pathway, including the JAK-STAT and ERK pathways, to promote cartilage regeneration.
  • Supports tissue repair processes such as bone healing and nerve regeneration.
  • Promotes cell proliferation, differentiation, and apoptosis.
  • Activates FGF/FR signaling pathways to regulate cartilage metabolism.
  • Exhibits high biological activity with an ED50 of <2 ng/mL.
  • Implicated in cancer research, particularly in glioblastoma multiforme (GBM).
  • Produced in E. coli with tag-free purification.