Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z030161
Encompass
Fibroblast Growth Factor-basic (FGF-basic) also known as HBGF-2 is a non-glycosylated heparin-binding growth factor that belongs to the FGF family. FGF-basic is present in basement membranes and in the subendothelial extracellular matrix of blood vessels. FGF-basic signals through FGFR1 2 3 and 4 that plays an important role in the regulation of cell survival cell division angiogenesis cell differentiation and cell migration.
Catalog No. 50-252-9884