Learn More
Genscript Corporation FGF-Basic Porcine
Catalog No. 5000057237
Encompass
Fibroblast Growth Factor-basic (FGF-basic) also known as FGF-2 is a pleiotropic cytokine FGF-basic is produced by fibroblasts and vascular cells It regulates a variety of processes including cell proliferation differentiation survival adhesion motility apoptosis limb formation and wound healing FGF-basic can be tumorigenic due to its role in angiogenesis and blood vessel remodeling
Catalog No. 50-000-57237 Supplier Genscript Corporation Supplier No. Z03822100
Fibroblast Growth Factor-basic (FGF-basic) also known as FGF-2 is a pleiotropic cytokine FGF-basic is produced by fibroblasts and vascular cells It regulates a variety of processes including cell proliferation differentiation survival adhesion motility apoptosis limb formation and wound healing FGF-basic can be tumorigenic due to its role in angiogenesis and blood vessel remodeling