Learn More
Genscript Corporation FGF-R4 Fc Chimera Mouse
Supplier:  Genscript Corporation Z035591
Encompass
Fibroblast growth factor receptor 4(FGFR4) also known as CD334 (cluster of differentiation 334) is a member of the fibroblast growth factor receptor family where amino acid sequence is highly conserved between members and throughout evolution A full-length representative protein would consist of an extracellular region composed of three immunoglobulin-like domains a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain The extracellular portion of the protein interacts with fibroblast growth factors setting in motion a cascade of downstream signals ultimately influencing mitogenesis and differentiation
Catalog No. 50-000-59359