Learn More
Aviva Systems Biology Corp FGF11 Antibody - N-terminal region (ARP54709_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54709_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGF11. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FGF11. Alternate names: FHF3, FLJ16061, MGC102953, MGC45269ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-958