Learn More
Aviva Systems Biology Corp FGF14 Antibody - middle region (ARP54710_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54710_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGF14. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FGF14. Alternate names: FHF4, MGC119129, SCA27, FHF-4, FGF-14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-959