Learn More
Biomatik Corporation Fgf15 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11086
Encompass_Preferred
Fgf15 Polyclonal Antibody is a Mus musculus (Mouse (Mus musculus)) Polyclonal Antibody to Fgf15. The Fgf15 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Mus musculus (Mouse (Mus musculus)). Immunogen: Recombinant Mouse Fibroblast Growth Factor 15 Protein (26-218Aa). Applications: ELISA, WB. Target Synonyms: Fgf15. Accession Number: O35622. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9252