Learn More
Aviva Systems Biology Corp FGF19 Antibody - C-terminal region (ARP63726_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63726_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGF19. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-288-065