Learn More
Biomatik Corporation Fgf19 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10725
Encompass_Preferred
Fgf19 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to FGF19. The Fgf19 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Fibroblast Growth Factor 19 Protein (25-216Aa). Applications: ELISA, WB, IHC. Target Synonyms: FGF19. Accession Number: O95750. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5099