Learn More
AdipoGen anti-FGF-19 (human), mAb (FG98-6) (50 µg)
SDP
Supplier:  AdipoGen AG20A0065C050
Encompass_Preferred
Monoclonal Antibody. Recognizes human FGF-19. Detects a band of ~25kDa by Western blot. Isotype: Mouse IgG2bkappa. Clone: FG98-6. Applications: ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. FGF-19 is involved in the suppression of bile acid biosynthesis through down-regulation of CYP7A1 expression, following positive regulation of the JNK and ERK1/2 cascades. Stimulates glucose uptake in adipocytes. FGF-19 plays an important role in brain development during embryogenesis. FGF-19 binds only to FGF receptor 4 (FGFR4). Activity requires the presence of KLB.
Catalog No. 50-205-2862