Learn More
Cayman Chemical FGF1humnrecombInt 100ug
SDP
Catalog No. 502949912
Encompass_Preferred
Source: Active recombinant human FGF1 expressed in E. coli • Amino acids: 16-155 • MW: 16 kDa
Catalog No. 50-294-9912 Supplier Cayman Chemical Supplier No. 33734100 UG
Source: Active recombinant human FGF1 expressed in E. coli • Amino acids: 16-155 • MW: 16 kDa