Learn More
Stemculture LLC StemCultures LLC FGF2 DISCs (Standard) – 10 ng/mL in 2 mL
SDP
Catalog No. NC2135529
Encompass
FGF2 DISCs are biocompatible hydrogels that release defined levels of native FGF2 stably into culture medium for one week. Native FGF2 has a short half-life (4 hours) and rapidly decays in cell culture media. Adding soluble FGF2 through daily media feeds creates fluctuations in growth factor levels that affect signaling pathways and creates a stressful cellular environment. With FGF2 DISCs, FGF2 levels are stable over one week, allowing the following reduced feeding with drastically improved cell quality and downstream applications. DISC device technology allows you to use your favorite media and feed less often. One DISC can be added per passage and can remain in the culture well/vessel for several media feeds (i.e., a pack of 48 DISCs will last 2 months when culturing in a 6 well plate). FGF2 DISCs are designed to release at a constant 10 ng/mL in 2 mL of medium. This technology gives scientists enhanced control over growth factor levels in cultures to provide high-quality cells.
This item is not returnable. View return policy
Catalog No. NC2135529 Supplier Stemculture LLC Supplier No. DSC50048
Add to Cart
Add to Cart
FGF2 DISCs are biocompatible hydrogels that release defined levels of native FGF2 stably into culture medium for one week. Native FGF2 has a short half-life (4 hours) and rapidly decays in cell culture media. Adding soluble FGF2 through daily media feeds creates fluctuations in growth factor levels that affect signaling pathways and creates a stressful cellular environment. With FGF2 DISCs, FGF2 levels are stable over one week, allowing the following reduced feeding with drastically improved cell quality and downstream applications. DISC device technology allows you to use your favorite media and feed less often. One DISC can be added per passage and can remain in the culture well/vessel for several media feeds (i.e., a pack of 48 DISCs will last 2 months when culturing in a 6 well plate). FGF2 DISCs are designed to release at a constant 10 ng/mL in 2 mL of medium. This technology gives scientists enhanced control over growth factor levels in cultures to provide high-quality cells.