Learn More
Aviva Systems Biology Corp FGF20 Antibody - C-terminal region (ARP64045_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64045_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGF20. It was validated on Western Blot. . Alternate names: FGF-20ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-154