Learn More
Genscript Corporation FGF21 mFc ChimeraBiotin Avi
Supplier:  Genscript Corporation Z04458500
Encompass
Fibroblast growth factor 21 (FGF21) is a peptide hormone that is synthesized by several organs and regulates energy homeostasis Excitement surrounding this relatively recently identified hormone is based on the documented metabolic beneficial effects of FGF21 which include weight loss and improved glycemia
Catalog No. 50-000-58175