Learn More
Genscript Corporation FGF21 mFc ChimeraBiotin Avi
Catalog No. 5000059182
Encompass
Fibroblast growth factor 21 (FGF21) is a peptide hormone that is synthesized by several organs and regulates energy homeostasis Excitement surrounding this relatively recently identified hormone is based on the documented metabolic beneficial effects of FGF21 which include weight loss and improved glycemia
Catalog No. 50-000-59182 Supplier Genscript Corporation Supplier No. Z04458100
Fibroblast growth factor 21 (FGF21) is a peptide hormone that is synthesized by several organs and regulates energy homeostasis Excitement surrounding this relatively recently identified hormone is based on the documented metabolic beneficial effects of FGF21 which include weight loss and improved glycemia