Learn More
Biotium FGF23(FGF23/638), CF740 conjugate, 0.1mg/mL
Supplier: Biotium BNC740638100
Fibroblast growth factor-1 (FGF-1, acidic FGF) and fibroblast growth factor-2 (FGF-2, basic FGF) are members of a family of growth factors that stimulate proliferation of cells of mesenchymal, epithelial and neuroectodermal origin.Additional members of the FGF family include the oncogenes FGF-3 (Int2) and FGF-4 (hst/Kaposi), FGF-5, FGF-6, FGF-7 (KGF), FGF-8 (AIGF), FGF-9 (GAF) and FGF-10 through FGF-23. Members of the FGF family share 30-55% amino acid sequence identity and similar gene structure. They are capable of transforming cultured cells when overexpressed by transfection. Cellular receptors for FGFs are members of a second multigene family, including four tyrosine kinases designated Flg (FGFR-1), Bek (FGFR-L), TKF and FGFR-3.Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommen