Learn More
AdipoGen anti-FGF-23 (human), mAb (FG322-3) (100 µg)
SDP
Supplier:  AdipoGen AG20A0073C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human FGF-23. Does not cross-react with mouse FGF-23. Isotype: Mouse IgG1kappa. Clone: FG322-3. Applications: ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. FGF-23 is a regulator of phosphate homeostasis. It upregulates EGR1 expression in the presence of KLBy. Acts directly on the parathyroid to decrease PTH secretion. Regulates the vitamin-D metabolism. Negatively regulates osteoblast differentiation and matrix mineralization. Defects in FGF-23 are the cause of autosomal dominant hypophosphataemic rickets (ADHR) and of hyperphosphatemic familial tumoral calcinosis (HFTC).
Catalog No. 50-205-2875