Learn More
ABclonal Technology FGF23 Rabbit mAb
SDP
Catalog No. 5000012796
Encompass_Preferred
This gene encodes a member of the fibroblast growth factor family of proteins which possess broad mitogenic and cell survival activities and are involved in a variety of biological processes The product of this gene regulates phosphate homeostasis and transport in the kidney The full-length functional protein may be deactivated via cleavage into N-terminal and C-terminal chains Mutation of this cleavage site causes autosomal dominant hypophosphatemic rickets (ADHR) Mutations in this gene are also associated with hyperphosphatemic familial tumoral calcinosis (HFTC)
Catalog No. 50-000-12796 Supplier ABclonal Technology Supplier No. A25006100UL
This gene encodes a member of the fibroblast growth factor family of proteins which possess broad mitogenic and cell survival activities and are involved in a variety of biological processes The product of this gene regulates phosphate homeostasis and transport in the kidney The full-length functional protein may be deactivated via cleavage into N-terminal and C-terminal chains Mutation of this cleavage site causes autosomal dominant hypophosphatemic rickets (ADHR) Mutations in this gene are also associated with hyperphosphatemic familial tumoral calcinosis (HFTC)