Learn More
Aviva Systems Biology Corp Fgf3 Antibody - C-terminal region (ARP42224_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42224_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fgf3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Int2, Fgf3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-284-985