Learn More
Enzo Life Sciences FGF7 (human), (recombinant) (10µg)
SDP
Catalog No. 502013992
Encompass_Preferred
Alternative name: KGF, Keratinocyte growth factor, Fibroblast growth factor 7. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from 1 mg/mL in 1X PBS. Source: Produced in HEK cells. Glycosylated protein. Long Term Storage: -20°C.
Catalog No. 50-201-3992 Supplier Enzo Life Sciences Supplier No. ENZPRT2020010
Alternative name: KGF, Keratinocyte growth factor, Fibroblast growth factor 7. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from 1 mg/mL in 1X PBS. Source: Produced in HEK cells. Glycosylated protein. Long Term Storage: -20°C.