Learn More
Aviva Systems Biology Corp FGF8 Antibody - C-terminal region (ARP63185_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63185_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGF8. It was validated on Western Blot. . Alternate names: AIGF, HBGF-8, KAL6, MGC149376, FGF-8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-904