Learn More
Sino Biological Fgf8 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 16124T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Fgf8 (rh Fgf8; Catalog#16124-HNAE; NP_149355.1; Gln23-Arg204). Fgf8 specific IgG was purified by Human Fgf8 affinity chromatography.
Catalog No. 50-224-7734