Learn More
ACROBiosystems Recombinant Protein;Human FGF-9 Protein, Fc Tag;HEK293;1MG;FG9,FGF9,FGF-9,GAF,HBFG-9,HBFG9,MGC119914,MGC119915,SYNS3
Supplier:  ACROBiosystems FG9H42671MG
Encompass_Preferred
Recombinant Protein;Human FGF-9 Protein, Fc Tag;1MG;Recombinant human FGF9 Fc chimera (rhFGF9-Fc) Leu4-Ser208 (Accession # AAI03979) was produced in human HEK293 cells at ACRObiosystems.;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 50.6 kDa. As a result of glycosylation, the protein migrates as 55 kDa under reducing (R) condition, and 110 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-7587