Learn More
Aviva Systems Biology Corp FGFBP2 Antibody - C-terminal region (ARP60593_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60593_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGFBP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human FGFBP2. Alternate names: HBP17RP, KSP37ApplicationWB. Species reactivity: Human
Catalog No. 50-287-097