Learn More
Medchemexpress LLC Fibroblast growth factor receptor 1 beta protein, human, hFc-tagged | 91.9% | 100UG
SDP
Catalog No. 5000177307
Encompass_Preferred
Recombinant human FGFR-1 beta extracellular domain fused to human Fc (hFc), expressed in HEK293 cells and supplied lyophilized for research use. The protein is formulated in 20 mM PB, 150 mM NaCl, pH 7.4, exhibits low endotoxin (<1 EU/μg), and shows bioactivity with an ED50 of 1.096 ng/mL in the presence of 1 ng/mL FGF-1. Reconstitute to ≥100 μg/mL and store aliquots at -20°C or -80°C.
  • Fc-tagged extracellular domain suitable for receptor binding and bioassays.
  • Expressed in HEK293 for human-like glycosylation and folding.
  • Lyophilized formulation for enhanced stability and transport.
  • Low endotoxin level supports cell-based assay use.
  • High specific activity (ED50 ~1.096 ng/mL) for reproducible results.
  • Clear reconstitution and storage guidance to preserve activity.
Catalog No. 50-001-77307 Supplier Medchemexpress LLC Supplier No. HYP700436100UG
Recombinant human FGFR-1 beta extracellular domain fused to human Fc (hFc), expressed in HEK293 cells and supplied lyophilized for research use. The protein is formulated in 20 mM PB, 150 mM NaCl, pH 7.4, exhibits low endotoxin (<1 EU/μg), and shows bioactivity with an ED50 of 1.096 ng/mL in the presence of 1 ng/mL FGF-1. Reconstitute to ≥100 μg/mL and store aliquots at -20°C or -80°C.
  • Fc-tagged extracellular domain suitable for receptor binding and bioassays.
  • Expressed in HEK293 for human-like glycosylation and folding.
  • Lyophilized formulation for enhanced stability and transport.
  • Low endotoxin level supports cell-based assay use.
  • High specific activity (ED50 ~1.096 ng/mL) for reproducible results.
  • Clear reconstitution and storage guidance to preserve activity.